siRNA Details
Gene Symbol: Coch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108710.1 | NP_001102180.1 | 4 | 175 | ||
GenBank mRNA | BC160874.1 | - | 146 | 7367398 | ||
FQ221314.1 | - | 232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108710.1 | NP_001102180.1 | 4 | 175 | ||
GenBank mRNA | BC160874.1 | - | 146 | 7367398 | ||
FQ221314.1 | - | 232 |