siRNA Details
Gene Symbol: LOC288978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134500.1 | NP_001127972.1 | 6 | 1029 | ||
XM_006249663.2 | XP_006249725.1 | 7 | 552 | |||
XM_008769430.1 | XP_008767652.1 | 6 | 667 |
