siRNA Details
Gene Symbol: LOC288978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134500.1 | NP_001127972.1 | 5, 6 | 959 | ||
XM_006249661.2 | XP_006249723.1 | 5, 6 | 597 | |||
XM_006249662.2 | XP_006249724.1 | 5, 6 | 597 | |||
XM_006249663.2 | XP_006249725.1 | 6, 7 | 482 | |||
XM_008769430.1 | XP_008767652.1 | 5, 6 | 597 | |||
XM_008769431.1 | XP_008767653.1 | 5, 6 | 597 |
