siRNA Details
Gene Symbol: Asun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109608.1 | NP_001103078.1 | 2 | 655 | ||
XM_006237689.1 | XP_006237751.1 | 2 | 700 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138757, BC027229, BY741532, BC026851, CF167981.
Gene Ontology Categories:
