siRNA Details
Gene Symbol: Gimap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034849.1 | NP_001030021.1 | 3 | 1035 | ||
XM_006236460.2 | XP_006236522.1 | 3 | 2441 | |||
XM_008762890.1 | XP_008761112.1 | 2 | 3126 | |||
XM_008762891.1 | XP_008761113.1 | 3 | 1867 | |||
GenBank mRNA | AJ633681.1 | - | 3 | 1521 | ||
BC127453.1 | - | 3 | 1029 | 7386337 | ||
CF108802.1 | - | 1 | 584 | |||
DQ125350.1 | - | 2 | 962 | |||
DQ125351.1 | - | 2 | 962 |
