siRNA Details
Gene Symbol: LOC683469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115040.1 | NP_001108512.1 | 3 | 1180 | ||
GenBank mRNA | BC158599.1 | - | 3 | 1180 | 7129113 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC683469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115040.1 | NP_001108512.1 | 3 | 1180 | ||
GenBank mRNA | BC158599.1 | - | 3 | 1180 | 7129113 |