siRNA Details
Gene Symbol: Dnajb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109541.1 | NP_001103011.1 | 4 | 396 | ||
XM_006245276.1 | XP_006245338.1 | 5 | 455 | |||
XM_006245278.2 | XP_006245340.1 | 5 | 477 | |||
XM_006245279.2 | XP_006245341.1 | 5 | 404 | |||
XM_006245280.1 | XP_006245342.1 | 5 | 455 | |||
XM_006245281.2 | XP_006245343.1 | 4 | 404 | |||
XM_006245283.1 | XP_006245345.1 | 5 | 455 | |||
GenBank mRNA | BC166710.1 | - | 5 | 392 | 7930287 |

More Information
Other Species Targeted

Gene Ontology Categories:
