siRNA Details
Gene Symbol: LOC689064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111269.1 | NP_001104739.1 | 578 | |||
GenBank mRNA | DY319996.1 | - | 578 | |||
FQ209804.1 | - | 598 | ||||
FQ209902.1 | - | 598 | ||||
FQ210004.1 | - | 595 | ||||
FQ210039.1 | - | 598 | ||||
FQ210298.1 | - | 596 | ||||
FQ210379.1 | - | 598 | ||||
FQ210599.1 | - | 596 | ||||
FQ210889.1 | - | 598 | ||||
FQ211252.1 | - | 596 | ||||
FQ211410.1 | - | 596 | ||||
FQ211709.1 | - | 671 | ||||
FQ215055.1 | - | 595 | ||||
FQ217121.1 | - | 627 | ||||
FQ217477.1 | - | 596 | ||||
FQ217746.1 | - | 596 | ||||
FQ218196.1 | - | 596 | ||||
FQ218287.1 | - | 596 | ||||
FQ221211.1 | - | 596 | ||||
FQ225430.1 | - | 596 | ||||
FQ225765.1 | - | 596 | ||||
FQ228130.1 | - | 1126 | ||||
FQ230470.1 | - | 598 | ||||
FQ230522.1 | - | 603 | ||||
FQ230899.1 | - | 600 | ||||
FQ231257.1 | - | 412 |
Gene Symbol: LOC103694857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773801.1 | XP_008772023.1 | 3 | 632 |
