siRNA Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033898.1 | NP_001029070.1 | 2 | 169 | ||
NM_022535.2 | NP_071980.2 | 2 | 169 | |||
GenBank mRNA | BC101908.1 | - | 2 | 169 | 7933832 | |
CK476875.1 | - | 2 | 190 | |||
FQ222101.1 | - | 1 | 12 |
