siRNA Details
Gene Symbol: Tmem86b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109604.1 | NP_001103074.1 | 3 | 520 | ||
XM_006228289.2 | XP_006228351.1 | 2 | 788 | |||
GenBank mRNA | BC158795.1 | - | 2 | 973 | 7314455 | |
FQ219460.1 | - | 3 | 434 |
