siRNA Details
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107107.1 | NP_001100577.2 | 10 | 3383 | ||
XM_006248177.2 | XP_006248239.1 | 10 | 3601 | |||
GenBank mRNA | FQ214009.1 | - | 1 | 570 |
