siRNA Details
Gene Symbol: Sestd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134514.2 | NP_001127986.2 | 18 | 2108 | ||
XM_006234402.2 | XP_006234464.1 | 19 | 2388 | |||
XM_006234404.2 | XP_006234466.1 | 18 | 2298 | |||
XM_008761892.1 | XP_008760114.1 | 19 | 2314 | |||
XM_008761893.1 | XP_008760115.1 | 18 | 2224 | |||
GenBank mRNA | BC169110.1 | - | 18 | 2108 | 7445081 |

More Information
Other Species Targeted

Gene Ontology Categories:
