siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 6 | 1415 | ||
NM_001199706.1 | NP_001186635.1 | 6 | 1648 | |||
NM_001199707.1 | NP_001186636.1 | 5 | 1656 | |||
NM_001199708.1 | NP_001186637.1 | 6 | 1489 | |||
NM_001300956.1 | NP_001287885.1 | 5 | 800 | |||
NM_015314.2 | NP_056129.2 | 5 | 1512 | |||
XM_005249689.3 | XP_005249746.1 | 6 | 1865 | |||
XM_005249690.4 | XP_005249747.1 | 5 | 962 | |||
GenBank mRNA | AB020702.1 | - | 5 | 1426 | ||
AK293912.1 | - | 6 | 1421 | |||
AK302559.1 | - | 6 | 1616 | |||
AK302632.1 | - | 4 | 592 | |||
BC028678.2 | - | 6 | 1374 | 4836984 | ||
BC056191.1 | - | 5 | 1396 | 5298453 | ||
BC136362.1 | - | 5 | 1406 | 9020349 | ||
BC144032.1 | - | 6 | 1553 | 9052552 | ||
BU507941.1 | - | 5 | 702 |
