siRNA Details
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134686.2 | NP_001128158.1 | 2 | 234 | ||
NM_022619.3 | NP_072141.2 | 2 | 234 | |||
XM_008771281.1 | XP_008769503.1 | 2 | 5313 | |||
XM_008771282.1 | XP_008769504.1 | 3 | 193 | |||
XM_008771283.1 | XP_008769505.1 | 2 | 5313 | |||
GenBank mRNA | AF245001.1 | - | 2 | 235 | ||
AF245002.1 | - | 2 | 235 |

More Information
Other Species Targeted

Gene Ontology Categories:
