siRNA Details
Gene Symbol: Rarg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135249.1 | NP_001128721.1 | 6 | 979 | ||
NM_001135250.1 | NP_001128722.1 | 4 | 751 | |||
XM_008765784.1 | XP_008764006.1 | 5 | 952 | |||
XM_008765785.1 | XP_008764007.1 | 6 | 943 | |||
XM_008765786.1 | XP_008764008.1 | 5 | 1471 |
