siRNA Details
Gene Symbol: Olr1537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001352.1 | NP_001001352.1 | 1 | 173 |
Gene Symbol: Olr1539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000729.1 | NP_001000729.1 | 173 |
