siRNA Details
Gene Symbol: Arvcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131013.1 | NP_001124485.1 | 7 | 1825 | ||
XM_006248617.2 | XP_006248679.1 | 5 | 4057 | |||
XM_006248618.2 | XP_006248680.1 | 5 | 4057 | |||
XM_008768880.1 | XP_008767102.1 | 7 | 1816 | |||
XM_008768881.1 | XP_008767103.1 | 6 | 2780 | |||
XM_008768882.1 | XP_008767104.1 | 6 | 1944 | |||
XM_008768883.1 | XP_008767105.1 | 6 | 2120 | |||
XM_008768884.1 | XP_008767106.1 | 6 | 2119 | |||
XM_008768885.1 | XP_008767107.1 | 5 | 4057 | |||
GenBank mRNA | BC168253.1 | - | 7 | 1825 | 5621230 |
