siRNA Details
Gene Symbol: Zfp949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134531.1 | NP_001128003.1 | 6 | 1989 | ||
XM_006243508.2 | XP_006243570.1 | 6 | 1983 | |||
XM_006243510.2 | XP_006243572.1 | 6 | 1926 | |||
XM_006243511.2 | XP_006243573.1 | 6 | 1471 |
