siRNA Details
Gene Symbol: Ormdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134704.1 | NP_001128176.1 | 200 | |||
GenBank mRNA | BC166795.1 | - | 200 | 7937873 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ormdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134704.1 | NP_001128176.1 | 200 | |||
GenBank mRNA | BC166795.1 | - | 200 | 7937873 |