siRNA Details
Gene Symbol: Atxn7l3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136261.1 | NP_001129733.1 | 1 | 3079 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atxn7l3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136261.1 | NP_001129733.1 | 1 | 3079 |