siRNA Details
Gene Symbol: LOC363337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014221.1 | NP_001014243.1 | 4, 5 | 1342 | ||
XM_006255363.2 | XP_006255425.1 | 5, 6 | 765 | |||
GenBank mRNA | BC082068.1 | - | 4, 5 | 1342 | 7132781 |
Gene Symbol: LOC102552540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255351.2 | XP_006255413.1 | 5, 6 | 883 |
Gene Symbol: RGD1562660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255382.2 | XP_006255444.1 | 5, 6 | 869 |
