siRNA Details
Gene Symbol: Eny2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130580.1 | NP_001124052.1 | 5 | 1797 | ||
XM_006241615.2 | XP_006241677.1 | 5 | 1763 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eny2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130580.1 | NP_001124052.1 | 5 | 1797 | ||
XM_006241615.2 | XP_006241677.1 | 5 | 1763 |