siRNA Details
Gene Symbol: Trip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134981.1 | NP_001128453.1 | 14 | 2376 | ||
XM_008766280.1 | XP_008764502.1 | 13 | 2183 | |||
XM_008766281.1 | XP_008764503.1 | 12 | 1844 | |||
XM_008766282.1 | XP_008764504.1 | 13 | 2306 | |||
XM_008766283.1 | XP_008764505.1 | 12 | 2408 | |||
XM_008766284.1 | XP_008764506.1 | 13 | 1975 | |||
GenBank mRNA | BC168748.1 | - | 13 | 1975 | 6921198 |
