siRNA Details
Gene Symbol: Oard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134596.1 | NP_001128068.1 | 2 | 475 | ||
XM_006244466.2 | XP_006244528.1 | 2 | 645 | |||
XM_006244467.2 | XP_006244529.1 | 2 | 642 | |||
XM_006244470.2 | XP_006244532.1 | 2 | 237 | |||
XM_006244471.2 | XP_006244533.1 | 2 | 470 | |||
XM_006244472.2 | XP_006244534.1 | 2 | 241 | |||
XM_008766872.1 | XP_008765094.1 | 2 | 653 | |||
XM_008766873.1 | XP_008765095.1 | 2 | 651 | |||
XM_008766874.1 | XP_008765096.1 | 3 | 517 | |||
XM_008766875.1 | XP_008765097.1 | 2 | 375 |
