siRNA Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047852.2 | NP_001041317.2 | 13 | 3056 | ||
XM_006227652.2 | XP_006227714.1 | 14 | 3223 | |||
XM_006227653.2 | XP_006227715.1 | 12 | 3052 | |||
XM_006227654.2 | XP_006227716.1 | 13 | 3080 | |||
XM_006227655.2 | XP_006227717.1 | 12 | 2939 | |||
XM_006227656.2 | XP_006227718.1 | 12 | 2933 | |||
XM_008758605.1 | XP_008756827.1 | 13 | 4215 | |||
GenBank mRNA | BC161968.1 | - | 13 | 3056 | 9028507 |

More Information
Other Species Targeted

Gene Ontology Categories:
