siRNA Details
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134854.1 | NP_001128326.1 | 4 | 1327 | ||
XM_006235063.2 | XP_006235125.1 | 5 | 1219 | |||
XM_006235064.2 | XP_006235126.1 | 5 | 1218 | |||
XM_006235065.2 | XP_006235127.1 | 5 | 1218 | |||
XM_008762165.1 | XP_008760387.1 | 5 | 1235 | |||
GenBank mRNA | BC160836.1 | - | 5 | 1102 | 7456805 | |
BC161894.1 | - | 4 | 1327 | 7193521 |
Gene Symbol: RGD1564480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_577020.6 | XP_577020.4 | 1 | 1118 |

More Information
Other Species Targeted

Gene Ontology Categories:
