siRNA Details
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134854.1 | NP_001128326.1 | 2 | 571 | ||
XM_006235063.2 | XP_006235125.1 | 3 | 463 | |||
XM_006235064.2 | XP_006235126.1 | 3 | 462 | |||
XM_006235065.2 | XP_006235127.1 | 3 | 462 | |||
XM_008762165.1 | XP_008760387.1 | 3 | 479 | |||
GenBank mRNA | BC160836.1 | - | 3 | 346 | 7456805 | |
BC161894.1 | - | 2 | 571 | 7193521 |
Gene Symbol: RGD1564480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773456.1 | XP_008771678.1 | 1 | 363 | ||
XM_577020.6 | XP_577020.4 | 1 | 362 |
