siRNA Details
Gene Symbol: NCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014286.3 | NP_055101.2 | 1, 2 | 150 | ||
GenBank mRNA | AF134479.1 | - | 1, 2 | 131 | ||
AF186409.1 | - | 1, 2 | 77 | |||
BC004856.2 | - | 1, 2 | 150 | 3928335 | ||
X84048.1 | - | 1, 2 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
