siRNA Details
Gene Symbol: LEPROTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015344.2 | NP_056159.2 | 4 | 447 | ||
GenBank mRNA | AF063605.1 | - | 4 | 424 | ||
AF161461.1 | - | 4 | 380 | |||
AK075363.1 | - | 4 | 391 | |||
AY277595.1 | - | 4 | 419 | |||
AY358674.1 | - | 4 | 419 | |||
BC000642.2 | - | 4 | 430 | 3344487 | ||
CR541735.1 | - | 3 | 340 | |||
CR541760.1 | - | 3 | 340 | |||
DA258969.1 | - | 4 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
