siRNA Details
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 15, 16 | 2903 | ||
XM_011536609.2 | XP_011534911.1 | 15, 16 | 2904 | |||
XM_017021124.1 | XP_016876613.1 | 15, 16 | 2904 | |||
XM_017021125.1 | XP_016876614.1 | 15, 16 | 2904 | |||
XM_017021126.1 | XP_016876615.1 | 14, 15 | 3518 | |||
XM_017021127.1 | XP_016876616.1 | 4, 5 | 600 | |||
XM_017021128.1 | XP_016876617.1 | 4, 5 | 453 | |||
GenBank mRNA | AB425197.1 | - | 15, 16 | 2891 | ||
BC144594.1 | - | 13, 14 | 2526 | |||
BX537886.1 | - | 15, 16 | 2897 | |||
BX538025.1 | - | 11, 12 | 2396 | |||
BX648683.1 | - | 15, 16 | 2901 | |||
CR749276.1 | - | 16, 17 | 2838 | |||
CR749365.1 | - | 15, 16 | 2902 |
