siRNA Details
Gene Symbol: GLTSCR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318819.1 | NP_001305748.1 | 14 | 3308 | ||
NM_015349.2 | NP_056164.1 | 12 | 3041 | |||
GenBank mRNA | AL833540.1 | - | 12 | 3041 | ||
BC047630.1 | - | 1 | 13 | |||
BC130302.1 | - | 12 | 2964 | 40146333 | ||
CR627378.1 | - | 13 | 3006 | |||
D87077.2 | - | 15 | 3205 |
