siRNA Details
Gene Symbol: CASP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136109.1 | NP_001129581.1 | 8 | 1119 | ||
NM_001136110.1 | NP_001129582.1 | 7 | 867 | |||
NM_001136112.1 | NP_001129584.1 | 9 | 1332 | |||
NM_004347.3 | NP_004338.3 | 9 | 1293 | |||
NR_024239.1 | 7 | 1009 | ||||
NR_036562.1 | 6 | 757 | ||||
GenBank mRNA | AK296660.1 | - | 8 | 1118 | ||
BC074994.2 | - | 8 | 1238 | 30915395 | ||
BC113406.1 | - | 8 | 1238 | 8322458 | ||
DQ228672.1 | - | 8 | 1261 | |||
DQ228673.1 | - | 7 | 1087 | |||
DQ228674.1 | - | 6 | 835 | |||
DQ228677.1 | - | 8 | 1300 | |||
U28015.1 | - | 8 | 1247 | |||
X94993.1 | - | 8 | 1154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
