siRNA Details
Gene Symbol: CASP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136112.1 | NP_001129584.1 | 2 | 213 | ||
NM_004347.3 | NP_004338.3 | 2 | 174 | |||
XM_011543021.2 | XP_011541323.1 | 2 | 908 | |||
GenBank mRNA | BC074994.2 | - | 1 | 119 | 30915395 | |
BC113406.1 | - | 1 | 119 | 8322458 | ||
DQ228672.1 | - | 1 | 142 | |||
DQ228677.1 | - | 1 | 181 | |||
DR005081.1 | - | 1 | 228 | |||
U28015.1 | - | 1 | 128 | |||
X94993.1 | - | 1 | 35 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
