siRNA Details
Gene Symbol: FLRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013280.4 | NP_037412.2 | 2 | 1882 | ||
XM_005273861.1 | XP_005273918.1 | 3 | 2768 | |||
XM_011544883.1 | XP_011543185.1 | 2 | 1722 | |||
GenBank mRNA | AF007139.1 | - | 1 | 257 | ||
AF169675.1 | - | 1 | 1541 | |||
AK289931.1 | - | 2 | 2199 | |||
BC018370.1 | - | 2 | 1882 | 4214494 |
