siRNA Details
Gene Symbol: NGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114090.1 | NP_001107562.1 | 13 | 1889 | ||
NM_019850.2 | NP_062824.2 | 15 | 2261 | |||
XM_011510923.2 | XP_011509225.1 | 15 | 2227 | |||
XM_011510924.1 | XP_011509226.1 | 11 | 1260 | |||
XM_011510925.1 | XP_011509227.1 | 11 | 1264 | |||
GenBank mRNA | AK293203.1 | - | 11 | 1261 | ||
AK297390.1 | - | 13 | 1889 | |||
AK300953.1 | - | 16 | 2165 | |||
BC031573.1 | - | 15 | 2248 | 5186132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
