siRNA Details
Gene Symbol: NGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019850.2 | NP_062824.2 | 2 | 459 | ||
XM_011510923.2 | XP_011509225.1 | 2 | 425 | |||
GenBank mRNA | AK298315.1 | - | 2 | 411 | ||
AK300953.1 | - | 3 | 363 | |||
BC031573.1 | - | 2 | 446 | 5186132 | ||
DB242442.1 | - | 2 | 459 | |||
DC388448.1 | - | 3 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
