siRNA Details
Gene Symbol: SLC39A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012319.3 | NP_036451.3 | 2 | 742 | ||
XM_011525900.1 | XP_011524202.1 | 2 | 1175 | |||
XM_011525901.1 | XP_011524203.1 | 3 | 1859 | |||
GenBank mRNA | AK304079.1 | - | 2 | 666 | ||
DC369482.1 | - | 1 | 227 | |||
U41060.2 | - | 3 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
