siRNA Details
Gene Symbol: RUNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001890.2 | NP_001001890.1 | 2 | 1961 | ||
NM_001122607.1 | NP_001116079.1 | 2 | 1961 | |||
NM_001754.4 | NP_001745.2 | 5 | 654 | |||
XM_005261068.3 | XP_005261125.1 | 3 | 917 | |||
XM_005261069.4 | XP_005261126.1 | 5 | 690 | |||
XM_011529766.2 | XP_011528068.1 | 5 | 765 | |||
XM_011529767.2 | XP_011528069.1 | 4 | 651 | |||
XM_011529768.2 | XP_011528070.1 | 4 | 651 | |||
XM_011529770.2 | XP_011528072.1 | 5 | 690 | |||
XM_017028487.1 | XP_016883976.1 | 3 | 459 | |||
GenBank mRNA | AK226159.1 | - | 4 | 651 | ||
AK310587.1 | - | 5 | 643 | |||
BC110828.1 | - | 4 | 549 | |||
BC136380.1 | - | 5 | 611 | 9020367 | ||
BC136381.1 | - | 5 | 610 | 9020368 | ||
BC144053.1 | - | 5 | 610 | 9052573 | ||
D10570.1 | - | 2 | 1151 | |||
D43967.1 | - | 2 | 1961 | |||
D43968.1 | - | 2 | 1961 | |||
D43969.1 | - | 4 | 908 | |||
D89788.1 | - | 3 | 351 | |||
D89789.1 | - | 3 | 351 | |||
D89790.1 | - | 3 | 351 | |||
L34598.1 | - | 2 | 1061 | |||
S60998.1 | - | 2 | 391 | |||
U19601.1 | - | 4 | 464 | |||
X79549.1 | - | 2 | 1951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:

Citations (1)
AML1 enhances the expression of leukotriene B4 type-1 receptor in leukocytes.
- Authors
- Hashidate T, Murakami N, Nakagawa M, Ichikawa M, Kurokawa M, Shimizu T, Nakamura M
- Journal
- Faseb J - Page No (Thu Apr 15 00:00:00 UTC 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
