siRNA Details
Gene Symbol: METTL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014033.3 | NP_054752.3 | 1 | 625 | ||
XM_006719332.1 | XP_006719395.1 | 2 | 979 | |||
GenBank mRNA | AF113007.1 | - | 1 | 299 | ||
AJ536233.1 | - | 1 | 347 | |||
AK023693.1 | - | 1 | 357 | |||
AK024409.1 | - | 1 | 390 | |||
AK225595.1 | - | 1 | 362 | |||
AK225689.1 | - | 1 | 361 | |||
AL050159.1 | - | 1 | 335 | |||
AY358797.1 | - | 1 | 393 | |||
BC004492.2 | - | 1 | 342 | 3689692 | ||
BC008180.2 | - | 1 | 343 | 2900478 | ||
BM805242.1 | - | 1 | 647 |
