siRNA Details
Gene Symbol: CNRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015463.2 | NP_056278.1 | 2, 3 | 979 | ||
XM_005264249.1 | XP_005264306.1 | 3, 4 | 674 | |||
XM_006711984.1 | XP_006712047.1 | 3, 4 | 442 | |||
GenBank mRNA | AK311784.1 | - | 2, 3 | 382 | ||
AL110235.1 | - | 2, 3 | 327 | |||
BC011535.1 | - | 2, 3 | 639 | 3870150 |

More Information
Other Species Targeted

