siRNA Details
Gene Symbol: CNRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015463.2 | NP_056278.1 | 3 | 1030 | ||
XM_005264249.1 | XP_005264306.1 | 4 | 725 | |||
XM_006711984.1 | XP_006712047.1 | 4 | 493 | |||
GenBank mRNA | AK092542.1 | - | 1 | 1671 | ||
AK311784.1 | - | 3 | 433 | |||
AL110235.1 | - | 3 | 378 | |||
BC011535.1 | - | 3 | 690 | 3870150 | ||
BC035125.1 | - | 1 | 3202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
