siRNA Details
Gene Symbol: ZNF385A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130967.2 | NP_001124439.1 | 4 | 337 | ||
NM_001130968.2 | NP_001124440.1 | 4 | 337 | |||
NM_001290001.1 | NP_001276930.1 | 3 | 432 | |||
NM_001290002.1 | NP_001276931.1 | 5 | 527 | |||
NM_001290004.1 | NP_001276933.1 | 5 | 482 | |||
NM_015481.2 | NP_056296.1 | 3 | 432 | |||
XM_005268783.4 | XP_005268840.1 | 4 | 359 | |||
XM_006719342.2 | XP_006719405.1 | 5 | 530 | |||
XM_011538168.2 | XP_011536470.1 | 5 | 449 | |||
XM_011538169.2 | XP_011536471.1 | 4 | 353 | |||
XM_011538170.1 | XP_011536472.1 | 3 | 202 | |||
XM_011538171.2 | XP_011536473.1 | 5 | 740 | |||
XM_017019174.1 | XP_016874663.1 | 4 | 665 | |||
GenBank mRNA | AB593085.1 | - | 5 | 392 | ||
AF304052.1 | - | 3 | 399 | |||
AK024404.1 | - | 4 | 313 | |||
AK092724.1 | - | 4 | 279 | |||
AK296564.1 | - | 3 | 375 | |||
AK315613.1 | - | 4 | 330 | |||
AL117462.1 | - | 2 | 42 | |||
AY461717.1 | - | 5 | 507 | |||
BC029752.1 | - | 4 | 318 | 4476647 | ||
BM546460.1 | - | 357 | ||||
BP305142.1 | - | 3 | 432 | |||
CR457327.1 | - | 3 | 276 | |||
DB116264.1 | - | 4 | 331 |
