siRNA Details
Gene Symbol: TMEM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286487.1 | NP_001273416.1 | 12 | 1275 | ||
NM_015497.4 | NP_056312.2 | 13 | 1336 | |||
XM_005254287.1 | XP_005254344.1 | 13 | 1190 | |||
XM_006720468.2 | XP_006720531.1 | 13 | 1195 | |||
XM_011521443.2 | XP_011519745.1 | 12 | 1107 | |||
XM_017022067.1 | XP_016877556.1 | 13 | 1192 | |||
GenBank mRNA | AF132733.1 | - | 13 | 1192 | ||
AK074870.1 | - | 13 | 1185 | |||
AK075403.1 | - | 13 | 1222 | |||
AK289591.1 | - | 13 | 1185 | |||
AK290150.1 | - | 13 | 1171 | |||
AK290484.1 | - | 12 | 1119 | |||
AK297816.1 | - | 13 | 1181 | |||
AK309915.1 | - | 13 | 1185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
