siRNA Details
Gene Symbol: C2CD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286577.1 | NP_001273506.1 | 2 | 541 | ||
NM_015531.5 | NP_056346.3 | 2 | 541 | |||
XM_011544904.1 | XP_011543206.1 | 2 | 440 | |||
XM_011544905.1 | XP_011543207.1 | 2 | 440 | |||
XM_017017510.1 | XP_016872999.1 | 2 | 440 | |||
XM_017017511.1 | XP_016873000.1 | 2 | 524 | |||
XM_017017512.1 | XP_016873001.1 | 2 | 479 | |||
XM_017017514.1 | XP_016873003.1 | 2 | 440 | |||
GenBank mRNA | AB231764.1 | - | 2 | 355 | ||
AK092089.1 | - | 2 | 479 | |||
BM460060.1 | - | 2 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
