siRNA Details
Gene Symbol: C2CD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286577.1 | NP_001273506.1 | 29 | 5782 | ||
NM_015531.5 | NP_056346.3 | 29 | 5782 | |||
XM_011544904.1 | XP_011543206.1 | 29 | 5681 | |||
XM_011544905.1 | XP_011543207.1 | 29 | 5681 | |||
XM_017017510.1 | XP_016872999.1 | 29 | 5681 | |||
XM_017017511.1 | XP_016873000.1 | 28 | 5607 | |||
XM_017017512.1 | XP_016873001.1 | 28 | 5572 | |||
XM_017017513.1 | XP_016873002.1 | 19 | 3778 | |||
XM_017017515.1 | XP_016873004.1 | 15 | 3331 | |||
XM_017017516.1 | XP_016873005.1 | 15 | 2901 | |||
GenBank mRNA | AB231764.1 | - | 29 | 5596 | ||
AK091397.1 | - | 13 | 2300 | |||
AK125894.1 | - | 16 | 3443 | |||
AL833903.1 | - | 10 | 1979 | |||
BC035599.1 | - | 9 | 1833 | |||
BC110508.2 | - | 16 | 3411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
