siRNA Details
Gene Symbol: NSMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130969.1 | NP_001124441.1 | 15, 16 | 1735 | ||
NM_001130970.1 | NP_001124442.1 | 14, 15 | 1666 | |||
NM_001130971.1 | NP_001124443.1 | 13, 14 | 1660 | |||
NM_001178064.1 | NP_001171535.1 | 14, 15 | 1645 | |||
NM_015537.4 | NP_056352.3 | 14, 15 | 1729 | |||
XM_005266061.4 | XP_005266118.1 | 13, 14 | 1662 | |||
XM_005266062.4 | XP_005266119.1 | 12, 13 | 1593 | |||
XM_011518496.2 | XP_011516798.1 | 13, 14 | 1599 | |||
XM_011518497.2 | XP_011516799.1 | 10, 11 | 849 | |||
XM_017014597.1 | XP_016870086.1 | 9, 10 | 1118 | |||
GenBank mRNA | AJ420418.1 | - | 7, 8 | 510 | ||
AK027474.1 | - | 12, 13 | 1000 | |||
AK074602.1 | - | 14, 15 | 1697 | |||
AK292030.1 | - | 10, 11 | 801 | |||
AK294320.1 | - | 10, 11 | 778 | |||
AK303976.1 | - | 10, 11 | 896 | |||
AK316046.1 | - | 12, 13 | 1051 | |||
AL117660.2 | - | 10, 11 | 757 | |||
AL389944.1 | - | 10, 11 | 783 | |||
AL389945.1 | - | 8, 9 | 1629 | |||
AL389946.1 | - | 6, 7 | 423 | |||
AY255128.1 | - | 15, 16 | 1735 | |||
AY255129.1 | - | 14, 15 | 1729 | |||
AY255130.1 | - | 14, 15 | 1666 | |||
AY255131.1 | - | 14, 15 | 1645 | |||
AY255132.1 | - | 14, 15 | 1971 | |||
BC004318.1 | - | 6, 7 | 403 | |||
BC016862.2 | - | 10, 11 | 776 | |||
BC072412.1 | - | 14, 15 | 1714 | 6165009 | ||
BC110498.2 | - | 13, 14 | 1527 | 40007676 |
