siRNA Details
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127231.2 | NP_001120703.1 | 17 | 3169 | ||
NM_015570.3 | NP_056385.1 | 18 | 3241 | |||
XM_005250257.2 | XP_005250314.1 | 11 | 1315 | |||
XM_011516010.2 | XP_011514312.1 | 18 | 3706 | |||
XM_011516011.2 | XP_011514313.1 | 18 | 3703 | |||
XM_011516012.2 | XP_011514314.1 | 17 | 3640 | |||
XM_011516013.2 | XP_011514315.1 | 17 | 3634 | |||
XM_011516014.2 | XP_011514316.1 | 16 | 3604 | |||
XM_011516017.2 | XP_011514319.1 | 17 | 12260 | |||
XM_011516018.2 | XP_011514320.1 | 18 | 2241 | |||
GenBank mRNA | AB007902.2 | - | 19 | 2730 | ||
AF326917.1 | - | 18 | 2827 | |||
AK296981.1 | - | 10 | 911 | |||
BC064693.1 | - | 17 | 3177 | 30345831 | ||
JQ670866.1 | - | 17 | 2434 | |||
JQ670867.1 | - | 9 | 838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
