siRNA Details
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 3 | 361 | ||
NM_001271869.1 | NP_001258798.1 | 3 | 358 | |||
NR_073501.1 | 4 | 432 | ||||
NR_073502.1 | 2 | 311 | ||||
NR_073503.1 | 2 | 308 | ||||
GenBank mRNA | AB097042.1 | - | 2 | 99 | ||
AB097043.1 | - | 2 | 99 | |||
AB097044.1 | - | 2 | 90 | |||
AB097045.1 | - | 2 | 90 | |||
AB097046.1 | - | 3 | 149 | |||
AJ295987.1 | - | 2 | 95 | |||
AK297926.1 | - | 3 | 149 | |||
AK298576.1 | - | 2 | 95 | |||
AK316205.1 | - | 2 | 97 | |||
AL050028.1 | - | 2 | 95 | |||
BC023988.1 | - | 2 | 116 | 3825796 | ||
BC065538.1 | - | 2 | 96 | 6053254 | ||
BX640985.1 | - | 2 | 111 | |||
DA010145.1 | - | 3 | 153 | |||
DA438597.1 | - | 4 | 219 | |||
DB501645.1 | - | 3 | 300 | |||
DC320437.1 | - | 2 | 313 |
Gene Symbol: LOC100130075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073494.1 | 1 | 139 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025608, NM_001277195, BC066992, AK027934, BB871722, and others.
