siRNA Details
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 5 | 1679 | ||
XM_005251439.4 | XP_005251496.1 | 5 | 1768 | |||
XM_006716754.3 | XP_006716817.1 | 3 | 234 | |||
XM_017014618.1 | XP_016870107.1 | 5 | 1885 | |||
XM_017014619.1 | XP_016870108.1 | 5 | 1996 | |||
GenBank mRNA | AB209610.1 | - | 4 | 1267 | ||
BC125124.1 | - | 3 | 273 | 40119018 | ||
BC125125.1 | - | 5 | 1758 | 40119019 | ||
BC140785.1 | - | 4 | 1650 | 9021653 | ||
BC171785.1 | - | 4 | 1650 | 9054439 |
