siRNA Details
Gene Symbol: FAM162A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014367.3 | NP_055182.3 | 5 | 476 | ||
GenBank mRNA | AF107495.1 | - | 5 | 457 | ||
AF191020.1 | - | 5 | 439 | |||
AF201944.1 | - | 5 | 469 | |||
AK026030.1 | - | 5 | 470 | |||
AK125835.1 | - | 1 | 4159 | |||
BC010896.1 | - | 5 | 474 | 4283379 | ||
BC015060.1 | - | 5 | 474 | 4248082 |
